Ward's® Prepared Potato Dextrose Agar
: Avantor
For growing bacteria and fungi
- Sterile- no autoclaving
- Convenient - prepared for you
- Prepared consistenly every time
Potato dextrose agar (or PDA agar) is a microbiological growth media made from potato infusion and dextrose.
: Some of these products contain living or perishable materials
: Prepared plates will ship via overnight or 2nd day delivery.
Prepared bottles and tubes do not require as much temperature regulation in-transit so we ship them via ground to save you the cost of freight. During checkout, please specify a delivery date for your specimens to arrive.
Prepared bottles and tubes do not require as much temperature regulation in-transit so we ship them via ground to save you the cost of freight. During checkout, please specify a delivery date for your specimens to arrive.
Frequently Bought Together