Potassium Bisulfate
Potassium Bisulfate, Reagent, 500 g
Supplier: Ward's Science
Fournisseur #: PP0370-500GR
Jump to:
Product Details & Documents
CAS Number: 7646-93-7
Formula Weight: 136.17
Formula: KHSO4
Density (g/mL): 2.245
Freezing Point (°C): 197
Solubility: Water
Synonyms: Potassium Hydrogen Sulfate
Shelf Life (months): 12
Storage: White
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Spécifications
Spécifications
- Désignation:Potassium Bisulfate
- Taille:500 g
- Chemical/Physical Specifications:Crystal
- Grade:Reagent
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



