Porcelain Spot Plates | Avantor
Porcelain Spot Plates
Porcelain Spot Plates
Certificates
Porcelain Spot Plates
Porcelain Spot Plates
2 Options Available Below
Product Details & Documents
These plates are for use in microchemistry experiments.
  • Glazed Porcelain
  • Acid-Resistant
  • Chemically-Inert
  • Plates Withstand up to 1000°F
The glazed porcelain spot plates are acid resistant and chemically inert.
They can withstand temperatures up to 1000°F.
Product Family Options
Informations sur le produit
  • Désignation
    Well Diameter
    Availability
    Price
  • VWR® Porcelain Spot Plates, 12 cavities, white
    22 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $38.55
    $38.55
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $437.25
    $437.25
    Each
       $38.55
    Case of 12
       $437.25
    Supplier Number: J-289
  • VWR® Porcelain Spot Plates, 30 cavities, white
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $159.50
    $159.50
    Each
       $159.50
    Supplier Number: J-291