Porcelain Buchner Funnels
Supplier: SILICO & CHEMICO PORCELAIN SE
Certificates
About this item
High Quality yet Affordable Buchner Funnels.
- Two sizes available
- Fragile
These high quality Buchner funnels are made from glazed perforated plate high-quality porcelain. They have a fixed, perforated plate and are glazed on the inside and outside with exception of the rim.
Caution: Fragile, use care when handling.
Frequently Bought Together