Pop Beads
: Avantor
Snap together beads.
- Multiple Colors Available
- Connectors Available
- Simulate Hydrogen Bonds with Special 5-Hole Beads
Build your own genetic models with these fun pop-beads. Specially designed white pop beads with four extra holes, equally spaced in a 90° plane from the axis of attachment, are typically used to form the nucleotide backbone. 10 mm diameter in size.
Pop Beads
Size: 10 mm dia. Packages of 200.
Pop Bead Connectors
Simulate hydrogen bonds. Package of 100.
Magnetic Centromeres
Package of 10.
Clear Centrioles
Package of 10.
: WARNING: CHOKING HAZARD. Children under 8 years can choke or suffocate. Adult supervision is required.
Magnetic Centromeres for Beads
Frequently Bought Together