Polypropylene Stirring Rod
Polypropylene Stirring Rod
Catalog # 470145-652
Supplier: PRECISION MOULDED POLYMERS LTD
: AM6010.00000.00000
Jump to:
Product Details & Documents
Made from high quality, unbreakable, 6 mm polypropylene tubing.
One end is square and the other end is diagonal. Size: 6 x 245mm.
Spécifications
- Désignation:Polypropylene Stirring Rod