ContactUsLinkComponent
Polypropylene Griffin Graduated Beakers
  470144-856
 : CH10155/1
undefined
Polypropylene Griffin Graduated Beakers
  470144-856
 :  CH10155/13
 : CH10155/1
Select an Option
5 ml
Select an Option
50 ml
Select an Option
Polypropylene Griffin Graduated Beakers
Select an Option
470144-856
Select an Option
BPO50
Economical, Unbreakable Beakers.
  • Permanent molded graduations
  • Special no-drip spouts
  • Autoclavable up to 121°C
Frequently Bought Together