Polypropylene Beaker Sets
Polypropylene Beaker Sets
 :   Dynalon
Polypropylene Beaker Sets
 :  Dynalon
Polypropylene Beaker Sets
Polypropylene Beaker Sets
Unbreakable Polypropylene
  • Autoclavable
  • Chemical-resistant
These beaker sets are autoclavable up to 145°C and chemical resistant. All feature pour spouts and metric graduation scales.
Product Family Options
  • Capacité
    Désignation
    Intervalle de graduation
  • 50 / 100 / 250 / 500 / 1000 ml
    Set of 5
    5 / 10 / 25 / 50 / 100 ml
    Kit of 5
    Kit of 5 - $90.90
    $90.90
    Kit of 5
       $90.90
     : 522135-0001
  • 250 / 500 / 1000 ml
    Set of 3
    25 / 50 / 100 ml
    Kit of 3
    Kit of 3 - $73.80
    $73.80
    Kit of 3
       $73.80
     : 522135-0003
  • 50 / 100 / 250 / 500 / 1000 ml
    Set of 5
    5 / 10 / 25 / 50 / 100 ml
     : 522135-0001