Polypropylene Aspirator Filter Pump | Avantor
Polypropylene Aspirator Filter Pump
Polypropylene Aspirator Filter Pump
Certificates
Polypropylene Aspirator Filter Pump
Polypropylene Aspirator Filter Pump
1 Options Available Below
Product Details & Documents
This lightweight polypropylene pump provides a constant vacuum when hooked to a standard faucet; it operates efficiently with water pressures as low as 7.5 psi for minimum water consumption. It has a maximum free-air pumping capacity of 11.5 liters per minute at a water-flow rate of 6.5 liters per minute. Ultimate vacuum: 28.5" Hg. Supplied with integral check valve, 3/8" NPT connecting thread, and 4 3/4" length of tubing for insertion in discharge end.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Aspirator Filter Pump
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $48.70
    $48.70
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $1,168.80
    $1,168.80
    Each
       $48.70
    Case of 24
       $1,168.80
    Supplier Number: 6140-0010
  • Aspirator Filter Pump
    Discontinued
    Supplier Number: 6140-0010