Polyethylene White Acid Cabinets
Supplier: Justrite
Certificates
About this item
Simple, Sleek Design for the Modern Lab
- Choose 1 or 2 manually closing door cabinet
- Door accepts padlock for security
- Feature Haz-Alert™ reflective warning labels
Solid polyethylene white acid cabinets have excellent chemical resistance to handle the most corrosive chemicals. A protective, all-welded bottom sump and a clean, sleek design complement any laboratory settings. Fully welded, liquid-tight 2" (51-mm) sump contains accidental spills and makes clean up a snap.
Ordering information: Ten year manufacturer warranty.
Frequently Bought Together