Justrite® Polyethylene Liquid Disposal Can
Justrite® Polyethylene Liquid Disposal Can
Polyethylene Liquid Disposal Can, 5 gal. (19 L), 20" H x 12" Outer Dia.
Catalog # 470202-912
Supplier:   Justrite
Supplier #:  12754
Justrite® Polyethylene Liquid Disposal Can
Catalog # 470202-912
Supplier:  Justrite
Justrite® Polyethylene Liquid Disposal Can
Justrite® Polyethylene Liquid Disposal Can
Polyethylene Liquid Disposal Can, 5 gal. (19 L), 20" H x 12" Outer Dia.
Price
$484.10
Log in pour la disponibilité
Each
Product Details & Documents
Superior Chemical Resistance and Leak Protection.
  • A safe solution for containing flammable wastes and corrosives
  • Constructed with high-density polyethylene
  • Controls vapors to reduce the risk of fire
Spring-loaded sealed cap, normally closed, automatically vents between 3 and 5 psig (0.2 and 0.35 bar). Stainless steel handle and hardware with latch-catch system hold the cap in the open position for easy filling and pouring. Large-mouth opening features a built-in stainless steel flame arrester to guard against outside ignition. The broad-base design provides stability and resists accidental tip over.
Spécifications
  • Capacité:
    5 gal. (19 L)
  • Désignation:
    Polyethylene Liquid Disposal Can
  • Exterior Dimensions:
    20" H x 12" Outer Dia.