Polyethylene Disposable Gloves
À propos de cet article
Ultra sensitive.
- Ambidextrous
- Powder-Free
These disposable, polyethylene gloves feature sheer-gauge 1.25 mil, which permits near bare hand sense of touch while protecting hands and labware from contamination. Gloves are powder-free, so they will not mark delicate materials.
3 Options Available Below
Product Details & Documents
Ultra sensitive.
- Ambidextrous
- Powder-Free
These disposable, polyethylene gloves feature sheer-gauge 1.25 mil, which permits near bare hand sense of touch while protecting hands and labware from contamination. Gloves are powder-free, so they will not mark delicate materials.
Specifications for Product Family
Spécifications
- Catalog No:
- 470016-978
- 470153-722
- 470153-724
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
Spécifications
Product Family Options
Informations sur le produit
- DésignationTaille du gantPackagingAvailabilityPrice
- Supplier Number: GDPEMDSpécifications:
More
Specifications
Cat. No.470016-978Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposablePowdered/Powder-freePowder-free - Polyethylene glovesMBox/100Box of 10018,00 $Case of 100001 604,80 $Supplier Number: GDPEMEDSpécifications:
More
Specifications
Cat. No.470153-722Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposablePowdered/Powder-freePowder-free - Polyethylene glovesLBox/100Box of 10022,50 $Case of 100001 995,20 $Supplier Number: GDPE-LGSpécifications:
More
Specifications
Cat. No.470153-724Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposablePowdered/Powder-freePowder-free