Polyester Seine
Polyester Seine, 10 ft., 4 ft., 1/4"
Catalog # 470018-572
Supplier: Peco Sales
Fournisseur #: 77699
Jump to:
Product Details & Documents
Great for Stream Sampling
- Large Area Screen for Sampling
- Collect More Fish and Specimens
It measures 10' long, 4' feet high, and is constructed of 1/4" polyester mesh. It comes with floats and leads, but not poles.
Spécifications
- Désignation:Polyester Seine
- Longueur:10 ft.
- Mesh Size:1/4"
- Width:4 ft.