Polycythemia Slide
Polycythemia Slide, Smear, Giemsa
Catalog # 470183-228
Supplier: Avantor
Jump to:
Product Details & Documents
Blood condition
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Peripheral blood. A chronic myeloproliferative disorder characterized by an increase in red blood cell mass, white blood cells, and platelets.
Spécifications
- Désignation:Polycythemia Slide
- Stain:Giemsa
- Orientation:Smear