Pocket Magnifiers
Pocket Magnifiers
Certificates
Pocket Magnifiers
Pocket Magnifiers
3 Options Available Below
Product Details & Documents
These pocket magnifiers are economical enough to outfit an entire class.
  • Handy plastic magnifiers
  • Ideal for younger students
  • Folds conveniently into attached case
  • Double/triple lenses can be stacked to increase magnification
Handy plastic magnifiers, with your choice of single, double, or triple lenses, are ideal for younger students. Each folds conveniently into an attached oval case. The 10X and 15X models’ double and triple lenses can be used separately or “stacked” to increase the magnification.
Dimensions: 1 1/2L x 1 1/4W x 1/4"H folded, 2 3/4"L extended.
Product Family Options
Informations sur le produit
  • Désignation
    Grossissement
    Availability
    Price
  • Pocket Magnifier Single Lens
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10.65
    $10.65
    Each
       $10.65
    Supplier Number: 23201-3
  • Pocket Magnifier Double Lens
    5×, 10×
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $21.65
    $21.65
    Log in pour la disponibilité
    Case of 72
    Case of 72 - $1,543.50
    $1,543.50
    Each
       $21.65
    Case of 72
       $1,543.50
    Supplier Number: 23202-3
  • Pocket Magnifier Triple Lens
    5×, 10×, 15×
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $27.55
    $27.55
    Each
       $27.55
    Supplier Number: 23203-3