Pocket Goniometer
Pocket Goniometer, 360°
Catalog # 470019-276
Supplier: Fabrication Enterprises
Fournisseur #: 12-1006
Jump to:
Product Details & Documents
Compact Size for Your Shirt Pocket
It is constructed of clear plastic and features a 5" linear scale calibrated in inches and centimeters.
Spécifications
- Désignation:Pocket Goniometer, 360°