Ward's® Pocket Gem Field Magnifier
Catalog # 470021-800
Supplier: COMPASS INDUSTRIES,INC.
Specifications
- Magnification:10×
- Description:Pocket Gem Field Magnifier
- Cat. no.:470021-800
- Supplier No.:792
Specifications
About this item
This doublet lens system at 10X provides sharp magnification.
- 10X magnification
This metal framed folding magnifier has a double lens system that provides 10X magnification. Size:1¼"Lx¾"Wx5/8"H folded,2¼"L extended. Lens size:½"dia.