Ward's® Pleochroism Slide Thin Section
 :   Avantor
Ward's® Pleochroism Slide Thin Section
 :  Avantor
Ward's® Pleochroism Slide Thin Section
Ward's® Pleochroism Slide Thin Section
Four mineral samples on one slide
  • A one of a kind slide
  • Created by Ward's geologists
  • Teach concepts that cannot be studied macroscopically
An innovative composite slide of four minerals displaying pleochroism; biotite, piedmontite, riebeckite, and tourmaline.
 : When minerals, rocks, and fossils cannot be accurately identified by macroscopic observation and testing, thin section slides can reveal the composition and structure of the specimens on a microscopic level. Thin section slides are precisely ground to a thickness of 0.03 mm across the entire specimen area. Mineral and rock thin sections are mounted on 27×46 mm microscope slides, and protected by a glass coverslip. All thin section slides are labeled for reference.
Product Family Options
  • Désignation
  • Pleochroism slide thin section
  • Pleochroism slide thin section
    Each
    Each - 236,98 $
    236,98 $
    Each
       236,98 $