Plastic Vials
Plastic Vials
Supplier:   Avantor
Plastic Vials
Supplier:  Avantor
Certificates
Plastic Vials
1 Options Available Below
Product Details & Documents
Made of safe, unbreakable plastic, the vials are available with or without friction-fit caps to prevent escape.
  • 3 oz. capacity
Use our foam plugs for ventilation. Size: 4×1 ³/₈".
Caution: Note: Not suitable for holding liquid.
Product Family Options
Product Information
  • Désignation
    Avec bouchon
    Availability
    Price
  • Vials, with caps
    Oui
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $26.85
    $26.85
    Pack of 10
       $26.85
  • Vials, without caps
    Non
    Discontinued