Plastic Trays
Plastic Trays, 450 x 350 x 75 mm (18 x 14 x 3")
Catalog # 470144-822
Supplier: United Scientific Supplies
Fournisseur #: 81701
Jump to:
Product Details & Documents
General use tray for all laboratory needs.
- Polypropylene
- Autoclavable
Multi-purpose polypropylene molded trays can be used for sterilizing, drying glassware, porcelain ware, and other laboratory supplies. These steam autoclavable trays have tapered walls designed for nesting.
Spécifications
- Désignation:Plastic Trays
- Dimensions:450 x 350 x 75 mm (18 x 14 x 3")