Plastic Protractor | Avantor
Plastic Protractor
Plastic Protractor
Catalog # 470151-092
Supplier:   Acme United
Fournisseur #:  376
Plastic Protractor
Catalog # 470151-092
Supplier:  Acme United
Plastic Protractor
Plastic Protractor
Plastic Protractor
Price
$5.30
Log in pour la disponibilité
Each Pack of 12 Case of 144
Product Details & Documents
Made of transparent plastic with scale printed on one side. Graduated 0 to 180° and 180 to 0° in 0.5° divisions. Diameter: 6".
Spécifications
  • Désignation:
    Plastic Protractor