Plastic Four-Sided Meter Stick
Metric meter stick, four-sided, Plastic, 1 m
Catalog # 470106-452
Supplier: SI Manufacturing
Supplier #: 45780-A
Jump to:
Product Details & Documents
Four meter sticks in one and made of flexible plastic.
The sides have 1 m, 10 dm, 100 cm, and 1000 mm graduations.
Specifications
- Désignation:Metric meter stick, four-sided
- Longueur:1 m
- Matière:Plastic