Plastic Chalkboard Compass | Avantor
Plastic Chalkboard Compass
Plastic chalkboard compass, 18"
Catalog # 470001-940
Fournisseur #:  CHCMP1
Plastic Chalkboard Compass
Catalog # 470001-940
Plastic Chalkboard Compass
Plastic Chalkboard Compass
Plastic chalkboard compass, 18"
Price
$70.15
Log in pour la disponibilité
Each
Product Details & Documents
Draw circles up to 50" in diameter.
  • Locking screw
  • Rubber suction cup
Draw circles up to a 50" diameter with this compass. One leg has suction cups for maintaining the center point on both black and white boards. The other leg has a spring tension holder for a pen, marker, or chalk.
Spécifications
  • Désignation:
    Plastic chalkboard compass, 18"