Plastic Back Thermometers | Avantor
Plastic Back Thermometers
Plastic Back Thermometers
Certificates
Plastic Back Thermometers
Plastic Back Thermometers
2 Options Available Below
Product Details & Documents
Great for Early Learning about Temperature Scales
  • Plastic shell provides better protection
  • Resistant to both chemical and heat
  • Choose between low range or high range thermometers
Teach students about temperature readings and the difference between the Celsius and Fahrenheit with these double scale thermometers.
Product Family Options
Informations sur le produit
  • Désignation
    Range
    Longueur
    Availability
    Price
  • Thermometer, Plastic Back, Low Range, Red Alcohol Filled
    −40…+50 °C (−40…+120 °F)
    14 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $12.65
    $12.65
    Each
       $12.65
    Supplier Number: AM1060.LB000.00000
  • Thermometer, Plastic Back, High Range, Red Alcohol Filled
    −30…+110 °C (−20…+230 °F)
    14 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $10.90
    $10.90
    Each
       $10.90
    Supplier Number: AM1060.HB000.00000