Plane Mirrors
Plane Mirrors
Plane Mirrors
Certificates
Plane Mirrors
5 Options Available Below
Product Details & Documents
All Types and Sizes for All Optical Applications
  • Front surface silvering eliminates double reflections
  • All produce virtual images
These plane mirrors are rectangular. They are available in glass or shatterproof Plexiglas. Back surface silvered mirrors are less expensive, and ideal for general use. The silvered back surface is painted for durability. Front surface silvering eliminates double reflections at the unsilvered surface, giving a brighter, sharper image. However, these mirrors are more easily damaged by scratching.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Glass mirror, front surface silvered, 50x50 mm
    Discontinued
    Supplier Number: CP32922-00
  • Plane Mirrors, Size, mm: 152 x 38 x 2.5
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $53.30
    $53.30
    Pack of 6
       $53.30
    Supplier Number: 2-393
  • Glass mirrors with seamed edges,152x100x6.3 mm
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $102.70
    $102.70
    Pack of 6
       $102.70
    Supplier Number: MSW6X4-PK/6
  • Plane Mirrors, Size, mm: 100 x 100 x 2.5
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $46.50
    $46.50
    Pack of 6
       $46.50
    Supplier Number: 1-980
  • Plexiglas mirrors, back surface silvered, 75x125x3 mm
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $28.20
    $28.20
    Pack of 6
       $28.20
    Supplier Number: QUOTE 3512N
  • Plexiglas mirrors, front surface silvered, 75x125x3 mm
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $48.65
    $48.65
    Pack of 6
       $48.65
    Supplier Number: 3512P