Pipette Support
Pipette Support
Catalog # 470145-334
Supplier: Bel-Art
Supplier #: F18953-0000_SE
Jump to:
Product Details & Documents
Sturdy molded polypropylene rack holds pipettes up to 1/2" in dia in 5 x 10 arrangement.
Lower deck keeps pipette tips from surface beneath. Convenient hand grips on both sides. Autoclavable at 121 °C.
Size: 8-3/4" H x 8-1/2" W x 4-1/2" D".
Specifications
- Désignation:Pipette Support