Pig Skull
Pig Skull, 1, 5", 10", 4.5"
Catalog # 470027-958
Supplier: Avantor
Jump to:
Product Details & Documents
Sus scrofa, the domestic pig, is an ideal representative of the pig family, Suidae. The skull is unmounted for closer examination.
- Natural bone
- Professionally cleaned and prepared
Ordering information: This is a real specimen, so size and appearance may vary from the image.
Spécifications
- Désignation:Pig Skull
- Hauteur:4.5"
- Longueur:10"
- Width:5"
- No. of Parts:1
- Cat. No.:470027-958