Phosphate, Low Range | Avantor
Phosphate, Low Range
Supplier:   LaMotte
Phosphate, Low Range
Supplier:  LaMotte
Certificates
Phosphate, Low Range
Phosphate, Low Range
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab. 
  • Range: 0–2 ppm
  • Material for 50 Tests
  • Compact, Waterproof Carrying Case Contains Kit
  • Instructions Included
  • Pre-calibrated Test Tubes
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Phosphate Refill Kit
    Discontinued
    Supplier Number: R-3121-01
  • LaMotte® Water Test Kit, Phosphate, Low Range
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $193.20
    $193.20
    Each
       $193.20
    Supplier Number: 3121-02