Phosphate Buffered Saline
Phosphate Buffered Saline
Supplier:   Ward's Science
Phosphate Buffered Saline
Supplier:  Ward's Science
Certificates
Phosphate Buffered Saline
Phosphate Buffered Saline
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Caution: Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Product Family Options
Product Information
  • Désignation
    Grade
    Chemical/Physical Specifications
    Taille
    Availability
    Price
  • Phosphate Buffered Saline, Solution
    Laboratory
    pH 7.2
    500 mL
    Log in for availability
    Log in for availability
    Each (500ml)
    Each (500ml) - $30.00
    $30.00
    Each (500ml)
       $30.00
    Supplier Number: PP0236-500ML
  • Phosphate Buffered Saline
    Laboratory
    Powder
    Makes 1 L of 10X
    Log in for availability
    Log in for availability
    Each (100g)
    Each (100g) - $22.20
    $22.20
    Each (100g)
       $22.20
    Supplier Number: PP0233-100G