Litmus Test Strips
Supplier: Avantor
Jump to:
À propos de cet article
No Color Chart Necessary.
- Pocket-sized, lightweight vial
- 100 litmus test strips per vial
3 Options Available Below
Product Details & Documents
No Color Chart Necessary.
- Pocket-sized, lightweight vial
- 100 litmus test strips per vial
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- pH test paper, blue litmus test (100 per vial, 6 vials)Log in pour la disponibilitéPack of 6$15.80