Permanent Markers
Black permanent markers
Catalog # 470153-994
Supplier: EATON OFFICE SUPPLY
Fournisseur #: ITA30016
Jump to:
Product Details & Documents
Mark any surface including glass, metal, or plastic with these fine felt-tip pens that use black instant-dry ink. Package of four.
Spécifications
- Désignation:Black permanent markers