Ward's® Peranema
Peranema (Small), Hay culture medium, 30, Jar
Catalog # 470177-302
Supplier: Ward's Science
Supplier #: PERNA30/35
Jump to:
Product Details & Documents
Extremely small euglenoid. Usually doesn’t rotate when swimming. Scavenger. Hay medium.
Characterized by flagella; lacks plastids.
Specifications
- Désignation:Peranema (Small)
- No. of Students:30
- Packaging:Jar
- Media:Hay culture medium