Ward's® Pegmatite
Ward's® Pegmatite
Supplier:   Avantor
Ward's® Pegmatite
Supplier:  Avantor
Certificates
Ward's® Pegmatite
Ward's® Pegmatite
3 Options Available Below
Product Details & Documents
Classic, very coarse-grained igneous texture.
  • Collected in South Dakotah
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Product Information
  • Désignation
    Specimen Dimensions
    Availability
    Price
  • Pegmatite, student specimens
    1×1 - 1×1"
    Log in for availability
    Log in for availability
    Pack of 10
    Pack of 10 - $31.45
    $31.45
    Pack of 10
       $31.45
  • Pegmatite, hand specimen
    2×3" - 3×4"
    Log in for availability
    Log in for availability
    Each
    Each - $32.35
    $32.35
    Each
       $32.35
  • Pegmatite, bulk pack, 1 kg
    Size will vary
    Log in for availability
    Log in for availability
    Each
    Each - $33.30
    $33.30
    Each
       $33.30
Storage
Product Information
  • Catalog #
    Désignation
    Availability
    Price
  • Storage box
    Log in for availability
    Log in for availability
    Each
    Each - $2.35
    $2.35
    Each
       $2.35