Pegasus Safety Spectacles
Pegasus safety spectacles
Supplier: VWR International
Jump to:
Product Details & Documents
Protect your eyes from debris.
- Eye protection for your educational and research needs
- Clear lenses are scratch resistant
The one-piece molded frame features side shields to protect the eyes from flying debris.
Certifications: ANSI Z87+ approved.
Spécifications
- Désignation:Pegasus safety spectacles