Peat Moss
Peat Moss
Catalog # 470153-626
Supplier: ARETT SALES CORP
Fournisseur #: H28-15503
Jump to:
Product Details & Documents
Mixed in soil, it improves moisture retention and aeration, as well as increases acidity. Use as a starter for seeds, rootings, transplants, and cuttings.
Quantity: 11 liters.
Spécifications
- Désignation:Peat Moss