Promega PCR Nucleotide Mix
Catalog # 470203-864
Supplier: PROMEGA SUNNYVALE INC SE
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Promega PCR Nucleotide Mix, 200 μL (40 mM)
- Cat. no.:470203-864
- Supplier No.:C1141
Specifications
About this item
10 mM concentration each of dATP, dCTP, dGTP, and dTTP, for a total nucleotide concentration of 40 mM.
Note: Can be stored at 4°C for up to six weeks, or -20°C for longer.