Patch Cord, Banana/Alligator
Catalog # 470020-588
Supplier: GSC International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Patch Cord, Banana/Alligator, 24"
- Cat. no.:470020-588
- Supplier No.:P56522
Specifications
About this item
Flexible, 18 gauge wire with a banana plug at one end and an alligator clip at the other.
The alligator clip has a protective vinyl sheath.
Packaging: Pack of six include three red and three black wires.