Paramecium caudatum Slide
Catalog # 470177-500
Supplier: Avantor
Specifications
- Description:Paramecium caudatum slide
- Cat. no.:470177-500
- Stain:Fast Green and Mallory's
- Supplier No.:WARD470177-500
- Orientation:Whole-mount
Specifications
About this item
Easy to see internal structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
Ideal for general study. Large cell with distinctive macro and micronucleus.