Parafilm
Parafilm
Parafilm
Certificates
Parafilm
2 Options Available Below
Product Details & Documents
Flexible, self-adhesive, waterproof plastic for closures, mounting and more.
Adheres quickly and firmly to itself, covering culture tube or flask mouths and sealing petri dish edges. Excellent resistance to air, other gases, alcohol, common acids, and moderately concentrated alkalies.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Parafilm, Roll Size, in. x ft.: 2 x 250
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $64.60
    $64.60
    Log in for availability
    Case of 24
    Case of 24 - $1,421.15
    $1,421.15
    Each
       $64.60
    Case of 24
       $1,421.15
    Supplier Number: HS234526A
  • Parafilm, Roll Size, in. x ft.: 20 x 50
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $125.55
    $125.55
    Log in for availability
    Case of 6
    Case of 6 - $715.35
    $715.35
    Each
       $125.55
    Case of 6
       $715.35
    Supplier Number: 120265