Overhead Flow Table | Avantor
Overhead Flow Table
Supplier:   DESK LABS
Overhead Flow Table
Supplier:  DESK LABS
Certificates
Overhead Flow Table
Overhead Flow Table
1 Options Available Below
Product Details & Documents
Demonstrate Earth Science Concepts in the Classroom
Simulate over 100 real-world water, air, and magma events through your overhead projector or hands-on labs. This kit includes 16 different plates that allow you to demonstrate everything from cloud formation to volcanic eruptions. This self-contained unit takes the place of a ripple tank, sand table or wave tank—allowing you to easily illustrate difficult earth and physical science concepts on the overhead. It can also be used as a wind tunnel.
This versatile demonstration is easy to clean and store.
Kit may be used on a classroom table, with an overhead projector or with a digital document camera projection system.
Ordering Information: Kit includes unit (16 x 16 x 2") chambered tray, water pump, tubing, fittings, event plates, eyedropper and teacher's guide with instructions. Kit does not include projection system, food coloring or rheoscopic fluid.
 : Kit includes unit (16 x 16 x 2") chambered tray, water pump, tubing, fittings, event plates, eyedropper and teacher's guide with instructions. Kit does not include projection system, food coloring or rheoscopic fluid.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Fluid Dynamics Simulator
    Discontinued
    Supplier Number: 1
  • Fluid Dynamics Simulator
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $221.95
    $221.95
    Kit of 1
       $221.95
    Supplier Number: 470200-900