Stage Micrometers
Stage Micrometer, Economy, —, 2 x 0.01 mm
Catalog # 470175-914
Supplier: Avantor
Jump to:
Product Details & Documents
Helpful microscope tool.
- Ruled 2 x 0.01 mm (wm) slide
- Shows 2 mm scale in 0.01 mm subdivisions
- Mounted on a regular glass slide
Use this stage micrometer to calibrate your microscope eyepiece reticles and objective powers.
Spécifications
- Désignation:Stage Micrometer, Economy
- Dimensions:2 x 0.01 mm
- Taille:—
Accessories
Informations sur le produit
- Catalog #No. of SlidesAvailabilityPrice
- Depth-of-field practice slide set
Informations sur le produit
- Catalog #AvailabilityPrice
- Eyepiece, DIN, 10× WF, w/Cross Hairs