Onchocerca volvulus, In Situ Slide
Onchocerca volvulus, In Situ, Section, Hematoxylin and Eosin
Catalog # 470182-156
Supplier: Avantor
Jump to:
Product Details & Documents
Species of Nematode
- Identifying features clearly distinguishable
- Rigorous quality control standards
Nodule showing adults packed with eggs and larvae. Causes onchocerciasis, fibrous skin tumors appearing as nodules. The organism may also be the agent of nodding syndrome.
Spécifications
- Désignation:Onchocerca volvulus, In Situ
- Stain:Hematoxylin and Eosin
- Orientation:Section