Ohaus® Replacement Hook Weights
Replacement Weight Hook, 100 g
Catalog # 470005-258
Supplier: OHAUS CORP.
Fournisseur #: 80850134
Jump to:
Product Details & Documents
Replacement hook and weights for Ohaus® products.
- Tamperproof
All Ohaus masses equal to or greater than 1 g are constructed of brass and are adjusted to the standards of the International Organization for Legal Metrology (OLM) M2 tolerance. Each mass is either one piece or drilled and then sealed, making the masses tamperproof.
Spécifications
- Désignation:Replacement Weight Hook
- Weight:100 g