Ward's® Obsidian
Ward's® Obsidian
Supplier:   Avantor
Ward's® Obsidian
Supplier:  Avantor
Certificates
Ward's® Obsidian
Ward's® Obsidian
4 Options Available Below
Product Details & Documents
Shiny black volcanic glass showing conchoidal fracture and some spherulites.
  • Collected in Mexico
  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available
Student Specimens – Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand Specimens – Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
Bulk Pack – Includes 1 kilo of assorted pieces (quantity varies). Assorted pieces are 1.25×1.25" to 2×3". Packaged in a labeled storage box for easy organization.
All specimen dimensions are considered an average size range.
Product Family Options
Specimens
Informations sur le produit
  • Désignation
    Specimen Dimensions
    Availability
    Price
  • Obsidian (thin section)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $59.19
    $59.19
    Each
       $59.19
  • Obsidian, student specimens
    1×1 - 1×1"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $26.70
    $26.70
    Pack of 10
       $26.70
  • Obsidian, hand specimen
    2×3" - 3×4"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $27.15
    $27.15
    Each
       $27.15
  • Obsidian, bulk pack, 1 kg
    Size will vary
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $34.01
    $34.01
    Each
       $34.01
Storage
Informations sur le produit
  • Catalog #
    Désignation
    Availability
    Price
  • Storage box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2.35
    $2.35
    Each
       $2.35