Obelia, Hydroids Slide
Obelia, Hydroids slide, Whole mount, Schneider's and Fast Green
Catalog # 470181-520
Supplier: Avantor
Jump to:
Product Details & Documents
Observe both reproductive and vegetative structures
- Identifying features clearly distinguishable
- Rigorous quality control standards
Hydroid colony showing reproductive and vegetative polyps.
Spécifications
- Désignation:Obelia, Hydroids slide
- Stain:Schneider's and Fast Green
- Orientation:Whole mount