Nylon Beaker Brushes | Avantor
Nylon Beaker Brushes
Supplier:   Justman Brush
Nylon Beaker Brushes
Supplier:  Justman Brush
Certificates
Nylon Beaker Brushes
2 Options Available Below
Product Details & Documents
For beakers, jars, cylinders, etc. With black nylon fibers and stiff tufted end on a wood handle.
Product Family Options
Informations sur le produit
  • Désignation
    Bristle Material
    Bristle Color
    Brush Head Length
    Ø tête de la brosse
    Handle Material
    Longueur totale
    Availability
    Price
  • Nylon Beaker Brush, Heavy Double Tufted End
    Nylon
    Black
    10.2 cm
    76.2 mm (3”)
    Wood
    30.5 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $76.40
    $76.40
    Each
       $76.40
    Supplier Number: J720392
  • Nylon Beaker Brush, Heavy Double Tufted End
    Nylon
    Black
    12.7 cm
    76.2 mm (3”)
    Wood
    40 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $74.60
    $74.60
    Each
       $74.60
    Supplier Number: J720400