Nutrient Broth
Nutrient Broth
Nutrient Broth, Laboratory, 454 g
Catalog # 470108-996
Supplier #:  CD146B
Nutrient Broth
Catalog # 470108-996
CAS Number:  
Nutrient Broth
Nutrient Broth
Nutrient Broth, Laboratory, 454 g
CAS Number:  
Price
$248.00
Log in for availability
Pack of 1
Product Details & Documents
General Purpose broth medium, includes label instructions for preparation. Must refrigerate upon receipt.
Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.
Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Specifications
Specifications
  • Désignation:
    Nutrient Broth
  • Taille:
    454 g
  • Chemical/Physical Specifications:
    Dehydrated
  • Grade:
    Laboratory
Specifications