Prepared Nutrient Agar | Avantor
Prepared Nutrient Agar
Prepared Nutrient Agar
Certificates
Prepared Nutrient Agar
Prepared Nutrient Agar
4 Options Available Below
Product Details & Documents
General-purpose media support the growth of a variety of bacteria and fungi.
  • Non-Selective, General-Purpose Media
  • Available as Slant Tubes, Petri Plates, and Heat-and-Pour Bottles
  • For Growing a Wide Range of Non-Fastidious Microorganisms
Nutrient agar, a non-selective media, is an excellent choice for growing a wide range of non-fastidious microorganisms. It is frequently used to isolate individual colonies from mixed cultures.
Tube media is configured as 12 tubes per pack. Petri dish media is configured as 10 plates per pack. Pour Paks come with three bottles of media (125 ml) and 20 sterile petri dishes. Just melt and pour.
Documents
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Type de conditionnement
    Availability
    Price
  • Nutrient Agar
    125 ml
    Bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $82.10
    $82.10
    Pack of 6
       $82.10
    Supplier Number: -
  • Nutrient Agar
    Plate
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $34.95
    $34.95
    Pack of 10
       $34.95
    Supplier Number: CM145-P25
  • Nutrient Agar
    Pour pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $42.60
    $42.60
    Each
       $42.60
    Supplier Number: -
  • Nutrient Agar
    12/pack
    Slant tube
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $52.10
    $52.10
    Pack of 12
       $52.10
    Supplier Number: 880490