Nucleic Acids Slide
Nucleic Acids, Section, Methyl Green and Pyronine
Catalog # 470182-574
Supplier: Avantor
Jump to:
Product Details & Documents
Investigate Genetic Material Under the Microscope
- Identifying features clearly distinguishable
- Rigorous quality control standards
Onion root tip. DNA stains blue; RNA stains red.
Spécifications
- Désignation:Nucleic Acids
- Stain:Methyl Green and Pyronine
- Orientation:Section