Nitrate-Nitrogen
About this item
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab.
- Range: 0 to 15 ppm
- Material for 50 tests
- Compact, waterproof carrying case contains kit
- Instructions included
- Pre-calibrated test tubes
- Aligned with GLOBE protocols
1 Options Available Below
Product Details & Documents
Pre-calibrated test tubes and labware give fast, accurate results in the classroom or in the lab.
- Range: 0 to 15 ppm
- Material for 50 tests
- Compact, waterproof carrying case contains kit
- Instructions included
- Pre-calibrated test tubes
- Aligned with GLOBE protocols
Product Family Options
Product Information
- DésignationAvailabilityPrice
- Nitrate Refill KitDiscontinued-Supplier Number: R-3354
- LaMotte® Water Test Kit, Nitrate-NitrogenDiscontinued-Supplier Number: 3354-01
- Supplier Number: 3354-CN-01